Boc-3-bromo-D-phenylalanine
Supplier: Matrix Scientific
Synonyms:
N-Boc-3-bromo-D-phenylalanine, (R)-3-(3-Bromophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid
041853-500MG
041853-1G
101853-466EA
304.62
USD
101853-466
101853-468
Boc-3-bromo-D-phenylalanine
Boc-3-bromo-D-phenylalanine
Hazard: Irritant
Formula:
C₁₄H₁₈BrNO₄ MW: 344.205 g/mol |
MDL Number:
MFCD01317704 CAS Number: 261360-77-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...