Boc-3-amino-L-tyrosine

Supplier: Matrix Scientific

041852-1G 041852-5G
101853-462EA 661.54 USD
101853-462 101853-464
Boc-3-amino-L-tyrosine
Boc-3-amino-L-tyrosine

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₅
MDL Number: MFCD02682254
CAS Number: 862191-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR