Boc-3,5-diiodo-L-tyrosine
Supplier: Matrix Scientific
Synonyms:
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid, Boc-Tyr(3,5-DiI)-OH, Boc-3,5-diiodo-Tyr-OH
041851-1G
041851-5G
101853-458EA
113.85
USD
101853-458
101853-460
Boc-3,5-diiodo-L-tyrosine
Boc-3,5-diiodo-L-tyrosine
Hazard: Irritant
Formula:
C₁₄H₁₇I₂NO₅ MW: 533.101 g/mol |
MDL Number:
MFCD00058515 CAS Number: 62129-53-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...