Boc-3,5-diiodo-L-thyronine
Supplier: Matrix Scientific
041850-500MG
041850-1G
101853-454EA
436.92
USD
101853-454
101853-456
Boc-3,5-diiodo-L-thyronine
Boc-3,5-diiodo-L-thyronine
Hazard: Irritant
Formula:
C₂₀H₂₁I₂NO₆ MW: 625.2 g/mol |
MDL Number:
MFCD02682264 CAS Number: 178877-78-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...