Boc-3,5-diiodo-D-tyrosine

Supplier: Matrix Scientific

Synonyms: Boc-3,5-diiodo-D-Tyr-OH

041849-1G 041849-5G
101853-450EA 498.46 USD
101853-450 101853-452
Boc-3,5-diiodo-D-tyrosine
Boc-3,5-diiodo-D-tyrosine

Hazard: Irritant

Formula: C₁₄H₁₇I₂NO₅
MW: 533.101 g/mol
MDL Number: MFCD00798624
CAS Number: 214630-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR