Boc-3,5,3#-triiodo-L-thyronine
Supplier: Matrix Scientific
041848-1G
041848-5G
101853-446EA
796.92
USD
101853-446
101853-448
Boc-3,5,3#-triiodo-L-thyronine
Boc-3,5,3#-triiodo-L-thyronine
Hazard: Irritant
Formula:
C₂₀H₂₀I₃NO₆ |
MDL Number:
MFCD02682295 CAS Number: 140396-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...