Boc-3,4-dihydroxy-L-phenylalanine

Supplier: Matrix Scientific

041846-1G 041846-5G
101853-438EA 430.77 USD
101853-438 101853-440
Boc-3,4-dihydroxy-L-phenylalanine
Boc-3,4-dihydroxy-L-phenylalanine

Hazard: Irritant

Formula: C₁₄H₁₉NO₆
MW: 297.31 g/mol
MDL Number: MFCD02682262
CAS Number: 30033-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR