Boc-3,4-dichloro-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: Boc-Phe(3,4-DiCl)-OH

041845-1G 041845-5G
101853-434EA 349.23 USD
101853-434 101853-436
Boc-3,4-dichloro-L-phenylalanine
Boc-3,4-dichloro-L-phenylalanine

Hazard: Irritant

Formula: C₁₄H₁₇Cl₂NO₄
MW: 334.2 g/mol
MDL Number: MFCD00273436
CAS Number: 80741-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR