Boc-3,4-dichloro-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: Boc-3,4-dichloro-D-Phe-OH, Boc-D-Phe(3,4-DiCl)-OH, Boc-D-Phe(3,4-Cl2)-OH

041844-1G 041844-5G
101853-430EA 176.92 USD
101853-430 101853-432
Boc-3,4-dichloro-D-phenylalanine
Boc-3,4-dichloro-D-phenylalanine

Hazard: Irritant

Formula: C₁₄H₁₇Cl₂NO₄
MW: 334.193 g/mol
MDL Number: MFCD00151887
CAS Number: 114873-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR