Boc-3,4,5-trifluoro-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-3-(3,4,5-trifluorophenyl)propanoic acid, 3,4,5-Trifluoro-L-phenylalanine, N-BOC protected

041843-1G 041843-5G
101853-426EA 498.46 USD
101853-426 101853-428
Boc-3,4,5-trifluoro-L-phenylalanine
Boc-3,4,5-trifluoro-L-phenylalanine

Hazard: Irritant

Formula: C₁₄H₁₆F₃NO₄
MW: 319.28 g/mol
MDL Number: MFCD00797558
CAS Number: 205445-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR