Boc-3,4,5-trifluoro-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)-3-(3,4,5-trifluorophenyl)propanoic acid, Boc-D-Phe(3,4,5-DiF)-OH

041842-1G 041842-5G
101853-422EA 498.46 USD
101853-422 101853-424
Boc-3,4,5-trifluoro-D-phenylalanine
Boc-3,4,5-trifluoro-D-phenylalanine

Hazard: Irritant

Formula: C₁₄H₁₆F₃NO₄
MW: 319.28 g/mol
MDL Number: MFCD00797559
CAS Number: 205445-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR