N-Boc-3-trifluoromethyl-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: Boc-Phe(3-CF3)-OH, (S)-2-((tert-Butoxycarbonyl)amino)-3-(3-(trifluoromethyl)phenyl)propanoic acid

041841-1G 041841-5G
101853-418EA 1095.38 USD
101853-418 101853-420
N-Boc-3-trifluoromethyl-L-phenylalanine
N-Boc-3-trifluoromethyl-L-phenylalanine

Hazard: Irritant

Formula: C₁₅H₁₈F₃NO₄
MW: 333.307 g/mol
MDL Number: MFCD00672524
CAS Number: 142995-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR