Boc-3-(trifluoromethyl)-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: Boc-D-Phe(3-CF3)-OH, Boc-D-Phe(3-CF₃)-OH

041840-1G 041840-5G
101853-414EA 526.15 USD
101853-414 101853-416
Boc-3-(trifluoromethyl)-D-phenylalanine
Boc-3-(trifluoromethyl)-D-phenylalanine

Hazard: Irritant

Formula: C₁₅H₁₈F₃NO₄
MW: 333.31 g/mol
MDL Number: MFCD00672525
CAS Number: 82317-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR