Boc-2-nitro-D-phenylalanine

Supplier: Matrix Scientific

041838-1G 041838-5G
101853-406EA 275.38 USD
101853-406 101853-408
Boc-2-nitro-D-phenylalanine
Boc-2-nitro-D-phenylalanine

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₆
MW: 310.31 g/mol
MDL Number: MFCD01317701
CAS Number: 478183-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR