Boc-2-methyl-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: Boc-D-Phe(2-Me)-OH

041836-1G 041836-5G
101853-398EA 164.62 USD
101853-398 101853-400
Boc-2-methyl-D-phenylalanine
Boc-2-methyl-D-phenylalanine

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MW: 279.34 g/mol
MDL Number: MFCD00671403
CAS Number: 80102-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR