Boc-D-2-cyanophenylalanine

Supplier: Matrix Scientific

Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)-3-(2-cyanophenyl)propanoic acid, N-Boc-2-cyano-D-phenylalanine

041834-1G 041834-5G
101853-390EA 547.69 USD
101853-390 101853-392
Boc-D-2-cyanophenylalanine
Boc-D-2-cyanophenylalanine

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₄
MW: 290.319 g/mol
MDL Number: MFCD01862940
CAS Number: 261380-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR