Boc-2-chloro-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: N-Boc-2-chloro-D-phenylalanine, (R)-2-((tert-Butoxycarbonyl)amino)-3-(2-chlorophenyl)propanoic acid

041832-1G 041832-5G
101853-382EA 138.46 USD
101853-382 101853-384
Boc-2-chloro-D-phenylalanine
Boc-2-chloro-D-phenylalanine

Hazard: Irritant

Formula: C₁₄H₁₈ClNO₄
MW: 299.751 g/mol
MDL Number: MFCD00801056
CAS Number: 80102-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR