N-Boc-2-bromo-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: Boc-D-Phe(2-Br)-OH

041830-1G 041830-5G
101853-374EA 412.31 USD
101853-374 101853-376
N-Boc-2-bromo-D-phenylalanine
N-Boc-2-bromo-D-phenylalanine

Hazard: Irritant

Formula: C₁₄H₁₈BrNO₄
MW: 344.205 g/mol
MDL Number: MFCD01317703
CAS Number: 261360-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR