Boc-2,4-dichloro-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-3-(2,4-dichlorophenyl)propanoic acid

041829-1G 041829-5G
101853-370EA 547.69 USD
101853-370 101853-372
Boc-2,4-dichloro-L-phenylalanine
Boc-2,4-dichloro-L-phenylalanine

Hazard: Irritant

Formula: C₁₄H₁₇Cl₂NO₄
MW: 334.2 g/mol
MDL Number: MFCD01862942
CAS Number: 114873-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR