N-Boc-2-trifluoromethyl-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: 2-(Trifluoromethyl)-D-phenylalanine, N-BOC protected, (R)-2-((tert-butoxycarbonyl)amino)-3-(2-(trifluoromethyl)phenyl)propanoic acid

041826-1G 041826-5G
101853-358EA 1095.38 USD
101853-358 101853-360
N-Boc-2-trifluoromethyl-D-phenylalanine
N-Boc-2-trifluoromethyl-D-phenylalanine

Hazard: Irritant

Formula: C₁₅H₁₈F₃NO₄
MW: 333.307 g/mol
MDL Number: MFCD01862945
CAS Number: 346694-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR