Boc-Tic-OH

Supplier: Matrix Scientific

Synonyms: (S)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, Boc-Tic-OH, (S)-2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, Boc-L-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, (S)-2-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

041825-1G 041825-5G
101853-354EA 15.38 USD
101853-354 101853-356
Boc-Tic-OH
Boc-Tic-OH

Hazard: Irritant

Formula: C₁₅H₁₉NO₄
MW: 277.32 g/mol
MDL Number: MFCD00143845
CAS Number: 78879-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR