N-BOC-erythro-L-β-Methylphenylalanine

Supplier: Matrix Scientific

Synonyms: (2S,3S)-Boc-β-methyl-L-Phe-OH

041823-250MG 041823-1G 041823-5G
101853-344EA 436.92 USD
101853-344 101853-346 101853-348
N-BOC-erythro-L-β-Methylphenylalanine
N-BOC-erythro-L-β-Methylphenylalanine

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MW: 279.332 g/mol
MDL Number: MFCD00800584
CAS Number: 90731-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR