(2R, 3S)/(2S, 3R)-racemic-Fmoc-β-hydroxyphenylalanine

Supplier: Matrix Scientific

041822-1G 041822-5G
101853-340EA 230.77 USD
101853-340 101853-342
(2R, 3S)/(2S, 3R)-racemic-Fmoc-β-hydroxyphenylalanine
(2R, 3S)/(2S, 3R)-racemic-Fmoc-beta-hydroxyphenylalanine

Hazard: Irritant

Formula: C₂₄H₂₁NO₅
MW: 403.44 g/mol
MDL Number: MFCD06656479
CAS Number: 487060-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR