(2R, 3S)/(2S, 3R)-racemic-Boc-β-hydroxyphenylalanine

Supplier: Matrix Scientific

041821-1G 041821-5G
101853-336EA 238.46 USD
101853-336 101853-338
(2R, 3S)/(2S, 3R)-racemic-Boc-β-hydroxyphenylalanine
(2R, 3S)/(2S, 3R)-racemic-Boc-beta-hydroxyphenylalanine

Hazard: Irritant

Formula: C₁₄H₁₉NO₅
MW: 281.31 g/mol
MDL Number: MFCD06656444
CAS Number: 102507-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR