(2R,3R)-Boc-β-methylphenylalanine

Supplier: Matrix Scientific

041820-250MG 041820-1G
101853-332EA 436.92 USD
101853-332 101853-334
(2R,3R)-Boc-β-methylphenylalanine
(2R,3R)-Boc-beta-methylphenylalanine

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MDL Number: MFCD00800585
CAS Number: 198493-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR