(2R,3R)/(2S,3S)-Racemic-boc-β-methylphenylalanine
Supplier: Matrix Scientific
Synonyms:
(2R, 3R)/(2S, 3S)-Racemic-Boc-beta-methyl-phenylalanine
041819-1G
041819-5G
101853-328EA
436.92
USD
101853-328
101853-330
(2R,3R)/(2S,3S)-Racemic-boc-β-methylphenylalanine
(2R,3R)/(2S,3S)-Racemic-boc-beta-methylphenylalanine
Hazard: Irritant
Formula:
C₁₅H₂₁NO₄ MW: 279.34 g/mol |
MDL Number:
MFCD01456490 CAS Number: 115132-19-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...