Fmoc-S-trityl-L-penicillamine

Supplier: Matrix Scientific

Synonyms: Fmoc-Pen(Trt)-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methyl-3-(tritylthio)butanoic acid

041818-1G 041818-5G
101853-324EA 461.54 USD
101853-324 101853-326
Fmoc-S-trityl-L-penicillamine
Fmoc-S-trityl-L-penicillamine

Hazard: Irritant

Formula: C₃₉H₃₅NO₄S
MW: 613.772 g/mol
MDL Number: MFCD00237388
CAS Number: 201531-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR