Fmoc-D-Pen(Trt)-OH

Supplier: Matrix Scientific

Synonyms: Fmoc-β,β-dimethyl-D-Cys(Trt)-OH, Fmoc-S-trityl-D-penicillamine

041817-1G 041817-5G
101853-320EA 461.54 USD
101853-320 101853-322
Fmoc-D-Pen(Trt)-OH
Fmoc-D-Pen(Trt)-OH

Hazard: Irritant

Formula: C₃₉H₃₅NO₄S
MW: 613.772 g/mol
MDL Number: MFCD00237389
CAS Number: 201532-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR