Fmoc-Cys(tBu)-OH

Supplier: Matrix Scientific

Synonyms: N-Fmoc-S-(tert-butyl)-L-cysteine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(tert-butyl)-L-cysteine, N-Fmoc-S-tert-butyl-L-cysteine, Fmoc-Cys(tBu)-OH

041816-5G 041816-25G
101853-316EA 224.62 USD
101853-316 101853-318
Fmoc-Cys(tBu)-OH
Fmoc-Cys(tBu)-OH

Hazard: Irritant

Formula: C₂₂H₂₅NO₄S
MDL Number: MFCD00037130
CAS Number: 67436-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR