Fmoc-Cys(Me)-OH

Supplier: Matrix Scientific

Synonyms: N-Fmoc-S-methyl-L-cysteine

041814-1G 041814-5G
101853-308EA 50.77 USD
101853-308 101853-310
Fmoc-Cys(Me)-OH
Fmoc-Cys(Me)-OH

Hazard: Irritant

Formula: C₁₉H₁₉NO₄S
MW: 357.424 g/mol
MDL Number: MFCD00237014
CAS Number: 138021-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR