Fmoc-Cys(Et)-OH

Supplier: Matrix Scientific

Synonyms: N-Fmoc-S-ethyl-L-cysteine

041813-1G 041813-5G
101853-304EA 138.46 USD
101853-304 101853-306
Fmoc-Cys(Et)-OH
Fmoc-Cys(Et)-OH

Hazard: Irritant

Formula: C₂₀H₂₁NO₄S
MW: 371.451 g/mol
MDL Number: MFCD00151921
CAS Number: 200354-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR