Fmoc-Cys(Dpm)-OH
Supplier: Matrix Scientific
041812-1G
041812-5G
101853-300EA
76.92
USD
101853-300
101853-302
Fmoc-Cys(Dpm)-OH
Fmoc-Cys(Dpm)-OH
Hazard: Irritant
Formula:
C₃₁H₂₇NO₄S MW: 509.63 g/mol Melting Pt: 73…78 °C |
MDL Number:
MFCD02682541 CAS Number: 247595-29-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...