Fmoc-S-carboxymethyl-L-cysteine
Supplier: Matrix Scientific
041811-1G
041811-5G
101853-296EA
275.38
USD
101853-296
101853-298
Fmoc-S-carboxymethyl-L-cysteine
Fmoc-S-carboxymethyl-L-cysteine
Hazard: Irritant
Formula:
C₂₀H₁₉NO₆S |
MDL Number:
MFCD02682534 CAS Number: 1354485-07-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...