Fmoc-S-carboxyethyl-L-cysteine

Supplier: Matrix Scientific

041810-1G 041810-5G
101853-292EA 326.15 USD
101853-292 101853-294
Fmoc-S-carboxyethyl-L-cysteine
Fmoc-S-carboxyethyl-L-cysteine

Hazard: Irritant

Formula: C₂₁H₂₁NO₆S
MW: 415.47 g/mol
MDL Number: MFCD02682532
CAS Number: 1354488-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR