Fmoc-S-carbamoyl-L-cysteine
Supplier: Matrix Scientific
041809-1G
041809-5G
101853-288EA
326.15
USD
101853-288
101853-290
Fmoc-S-carbamoyl-L-cysteine
Fmoc-S-carbamoyl-L-cysteine
Hazard: Irritant
Formula:
C₁₉H₁₈N₂O₅S |
MDL Number:
MFCD02682530 CAS Number: 1354488-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...