Fmoc-cys(Bzl)-OH

Supplier: Matrix Scientific

Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(benzylthio)propanoic acid, N-Fmoc-S-benzyl-L-cysteine

041808-5G 041808-25G
101853-284EA 175.38 USD
101853-284 101853-286
Fmoc-cys(Bzl)-OH
Fmoc-cys(Bzl)-OH

Hazard: Irritant

Formula: C₂₅H₂₃NO₄S
MW: 433.522 g/mol
MDL Number: MFCD00065633
CAS Number: 53298-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR