Fmoc-S-benzyl-DL-homocysteine

Supplier: Matrix Scientific

041807-250MG 041807-1G 041807-5G
101853-278EA 498.46 USD
101853-278 101853-280 101853-282
Fmoc-S-benzyl-DL-homocysteine
Fmoc-S-benzyl-DL-homocysteine

Hazard: Irritant

Formula: C₂₆H₂₅NO₄S
MDL Number: MFCD02682517
CAS Number: 1452573-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR