Fmoc-D-Cys(Bzl)-OH

Supplier: Matrix Scientific

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(benzylthio)propanoic acid

041806-1G 041806-5G
101853-274EA 249.23 USD
101853-274 101853-276
Fmoc-D-Cys(Bzl)-OH
Fmoc-D-Cys(Bzl)-OH

Hazard: Irritant

Formula: C₂₅H₂₃NO₄S
MW: 433.522 g/mol
MDL Number: MFCD02259496
CAS Number: 252049-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR