Fmoc-Pen(Acm)-OH

Supplier: Matrix Scientific

041805-1G 041805-5G
101853-270EA 412.31 USD
101853-270 101853-272
Fmoc-Pen(Acm)-OH
Fmoc-Pen(Acm)-OH

Hazard: Irritant

Formula: C₂₃H₂₆N₂O₅S
MW: 442.53 g/mol
MDL Number: MFCD00151935
CAS Number: 201531-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR