Fmoc-D-Pen(Acm)-OH
Supplier: Matrix Scientific
041804-1G
041804-5G
101853-266EA
412.31
USD
101853-266
101853-268
Fmoc-D-Pen(Acm)-OH
Fmoc-D-Pen(Acm)-OH
Hazard: Irritant
Formula:
C₂₃H₂₆N₂O₅S |
MDL Number:
MFCD00151934 CAS Number: 201531-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...