Fmoc-D-Pen(pMeOBzl)-OH

Supplier: Matrix Scientific

041802-1G 041802-5G
101853-258EA 363.08 USD
101853-258 101853-260
Fmoc-D-Pen(pMeOBzl)-OH
Fmoc-D-Pen(pMeOBzl)-OH

Hazard: Irritant

Formula: C₂₈H₂₉NO₅S
MDL Number: MFCD02094102
CAS Number: 159618-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR