Fmoc-S-[2-(4-pyridyl)ethyl]-DL-penicillamine

Supplier: Matrix Scientific

041800-100MG 041800-250MG
101853-250EA 412.31 USD
101853-250 101853-252
Fmoc-S-[2-(4-pyridyl)ethyl]-DL-penicillamine
Fmoc-S-[2-(4-pyridyl)ethyl]-DL-penicillamine

Hazard: Irritant

Formula: C₂₇H₂₈N₂O₄S
MDL Number: MFCD02682565
CAS Number: 1219404-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR