Fmoc-L-selenomethionine

Supplier: Matrix Scientific

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(methylselanyl)butanoic acid, Fmoc-SeMet-OH

041799-500MG 041799-1G
101853-246EA 430.77 USD
101853-246 101853-248
Fmoc-L-selenomethionine
Fmoc-L-selenomethionine

Hazard: Irritant/Store in Freezer

Formula: C₂₀H₂₁NO₄Se
MW: 418.35 g/mol
MDL Number: MFCD03093662
CAS Number: 1217852-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR