Fmoc-2-amino-4-(ethylthio)butyric acid

Supplier: Matrix Scientific

041798-250MG 041798-1G 041798-5G
101853-240EA 300 USD
101853-240 101853-242 101853-244
Fmoc-2-amino-4-(ethylthio)butyric acid
Fmoc-2-amino-4-(ethylthio)butyric acid

Hazard: Irritant

Formula: C₂₁H₂₃NO₄S
MDL Number: MFCD01861306
CAS Number: 1562431-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR