Fmoc-DL-selenomethionine
Supplier: Matrix Scientific
041795-500MG
041795-1G
101853-226EA
823.08
USD
101853-226
101853-228
Fmoc-DL-selenomethionine
Fmoc-DL-selenomethionine
Hazard: Irritant
Formula:
C₂₀H₂₁NO₄Se MW: 418.35 g/mol |
MDL Number:
MFCD02682569 CAS Number: 1219375-33-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...