Fmoc-DL-ethionine

Supplier: Matrix Scientific

041793-1G 041793-5G
101853-218EA 498.46 USD
101853-218 101853-220
Fmoc-DL-ethionine
Fmoc-DL-ethionine

Hazard: Irritant

Formula: C₂₁H₂₃NO₄S
MDL Number: MFCD09750516
CAS Number: 1396988-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR