Fmoc-DL-buthioninesulfoximine

Supplier: Matrix Scientific

041792-250MG 041792-1G 041792-5G
101853-212EA 275.38 USD
101853-212 101853-214 101853-216
Fmoc-DL-buthioninesulfoximine
Fmoc-DL-buthioninesulfoximine

Hazard: Irritant

Formula: C₂₃H₂₈N₂O₅S
MW: 444.55 g/mol
MDL Number: MFCD02682525
CAS Number: 1396987-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR