Fmoc-DL-buthionine

Supplier: Matrix Scientific

041791-1G 041791-5G
101853-208EA 661.54 USD
101853-208 101853-210
Fmoc-DL-buthionine
Fmoc-DL-buthionine

Hazard: Irritant

Formula: C₂₃H₂₇NO₄S
MDL Number: MFCD12547718
CAS Number: 1396969-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR