Fmoc-D-2-amino-4-(ethylthio)butyric acid

Supplier: Matrix Scientific

041790-250MG 041790-1G 041790-5G
101853-202EA 300 USD
101853-202 101853-204 101853-206
Fmoc-D-2-amino-4-(ethylthio)butyric acid
Fmoc-D-2-amino-4-(ethylthio)butyric acid

Hazard: Irritant

Formula: C₂₁H₂₃NO₄S
MDL Number: MFCD01861364
CAS Number: 1374669-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR