Fmoc-α-methyl-DL-methionine

Supplier: Matrix Scientific

041787-1G 041787-5G
101853-186EA 958.46 USD
101853-186 101853-188
Fmoc-α-methyl-DL-methionine
Fmoc-alpha-methyl-DL-methionine

Hazard: Irritant

Formula: C₂₁H₂₃NO₄S
MW: 385.49 g/mol
MDL Number: MFCD02682557
CAS Number: 147108-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR